Learn More
Apexbio Technology LLC 2'-O-Methyl-CTP(Synonyms: 2'-O-Methylcytidine-5'-triphosphate, 2'-OMe-CTP, 2'-O-Methyl CTP, 2'-Methoxy CTP), 50ul (100 mM).
Supplier: Apexbio Technology LLC B79690.05
2 -O-Methyl-CTP is a chemically modified analog of cytidine triphosphate (CTP) characterized by methyl substitution at the 2 -hydroxyl position of the ribose sugar This modification confers increased resistance against nuclease-mediated degradation when incorporated into RNA transcripts enhancing RNA stability It is commonly employed in biomedical research for investigating RNA structure-function relationships assessing RNA stability and synthesizing siRNA or miRNA analogs utilized in RNA interference (RNAi) studies