Learn More
Cayman Chemical 2-O-methyl PAF C-16, Purity: ≥98%, CAS: 78858-44-3, 25 mg
SDP
Catalog No. 502893719
Encompass_Preferred
2-O-methyl PAF C-16 is a synthetic PAF analog which contains a methyl group, attached by an ether linkage, at the sn-2 position. Although the biological activities of 2-O-methyl PAF C-16 have not been well characterized, the C-18 analog has been shown to mediate a number of biological responses including decreasing plasma membrane fluidity and inhibiting tumor cell invasiveness in embryonic chick heart.{2099} In rat astrocytes, the C-18 analog causes the release of large quantities of NO by a pathway involving activation of NOS.{2985}
Catalog No. 50-289-3719 Supplier Cayman Chemical Supplier No. 6090225 MG
2-O-methyl PAF C-16 is a synthetic PAF analog which contains a methyl group, attached by an ether linkage, at the sn-2 position. Although the biological activities of 2-O-methyl PAF C-16 have not been well characterized, the C-18 analog has been shown to mediate a number of biological responses including decreasing plasma membrane fluidity and inhibiting tumor cell invasiveness in embryonic chick heart.{2099} In rat astrocytes, the C-18 analog causes the release of large quantities of NO by a pathway involving activation of NOS.{2985}