Learn More
Apexbio Technology LLC 2'-O-Methyl-UTP(Synonyms: 2'-OMe-UTP, 2'-O-Methyluridine-5'-triphosphate, 2'-Methoxyuridine-5'-triphosphate), 10ul (100 mM).
SDP
Supplier:  Apexbio Technology LLC B79710.01
Encompass_Preferred
2 -O-Methyl-UTP is a chemically modified analog of uridine triphosphate (UTP) in which the 2 -hydroxyl group of the ribose sugar is replaced by a methyl group This structural alteration confers enhanced resistance to nuclease degradation thereby improving the stability of RNA molecules Due to its heightened stability 2 -O-Methyl-UTP is frequently incorporated into RNA oligonucleotides for RNA interference (RNAi) antisense oligonucleotide research and mRNA modification studies Its application extends broadly to investigations involving gene expression regulation RNA therapeutic designs and nucleic acid delivery platforms in molecular biology and biomedicine
Catalog No. 50-000-60262