Learn More
Cayman Chemical 2'-O-Methyladenosine, Purity: ≥98%, CAS: 2140-79-6, 50 mg
SDP
Catalog No. 502967578
Encompass_Preferred
An analog of adenosine used to prepare nucleoside derivatives as inhibitors of viral RNA translation and replication
Catalog No. 50-296-7578 Supplier Cayman Chemical Supplier No. 1693650 MG
An analog of adenosine used to prepare nucleoside derivatives as inhibitors of viral RNA translation and replication