Learn More
Medchemexpress LLC HY-W011552 10mg Medchemexpress, 2'-O-Methyladenosine CAS:2140-79-6 Purity:>98%
SDP
Catalog No. 501970683
Encompass_Preferred
Medchemexpress, HY-W011552 10mg 2'-O-Methyladenosine CAS:2140-79-6 2'-O-Methyladenosine, a methylated adenine residue is found in urine of normals as well as in urine of adenosine deaminase (ADA) deficient patients. 2'-O-Methyladenosine exhibits unique hypotensive activities . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0683 Supplier Medchemexpress LLC Supplier No. HYW01155210MG
Medchemexpress, HY-W011552 10mg 2'-O-Methyladenosine CAS:2140-79-6 2'-O-Methyladenosine, a methylated adenine residue is found in urine of normals as well as in urine of adenosine deaminase (ADA) deficient patients. 2'-O-Methyladenosine exhibits unique hypotensive activities . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.