Learn More
Medchemexpress LLC 2'-O-Methyladenosine | 2140-79-6 | 99.8% | 281.27 | 100 MG
SDP
Catalog No. 5000455091
Encompass_Preferred
2'-O-Methyladenosine is a methylated adenine residue found in the urine of both normal individuals and adenosine deaminase (ADA) deficient patients. It exhibits unique hypotensive activities.
  • Human endogenous metabolite.
  • Exhibits unique hypotensive activities in rats.
  • Decreased blood pressure by about 40-50% in male spontaneously hypertensive rats.
Catalog No. 50-004-55091 Supplier Medchemexpress LLC Supplier No. HYW011552100MG
2'-O-Methyladenosine is a methylated adenine residue found in the urine of both normal individuals and adenosine deaminase (ADA) deficient patients. It exhibits unique hypotensive activities.
  • Human endogenous metabolite.
  • Exhibits unique hypotensive activities in rats.
  • Decreased blood pressure by about 40-50% in male spontaneously hypertensive rats.