Learn More
Cayman Chemical 2′-O-Methylcytidine, Purity: ≥98%, CAS: 2140-72-9, 1 g
SDP
Catalog No. 502950289
Encompass_Preferred
An inhibitor of HCV NS5B (IC50 = 3.8 µM); inhibits HCV replication in a replicon assay (IC50 = 21.2 µM)
Catalog No. 50-295-0289 Supplier Cayman Chemical Supplier No. 334111 G
An inhibitor of HCV NS5B (IC50 = 3.8 µM); inhibits HCV replication in a replicon assay (IC50 = 21.2 µM)