Learn More
Medchemexpress LLC 2'-O-Methylcytidine | 2140-72-9 | 99.7% | 257.25 | 1 G
SDP
Catalog No. 5000452584
Encompass_Preferred
2'-O-Methylcytidine is an orally active 2'-substituted nucleoside that acts as an inhibitor of HCV replication with antiviral activity. It inhibits RNA-dependent RNA polymerase (NS5B)-catalyzed RNA synthesis in vitro by competing with substrate nucleoside triphosphate.
  • Orally active 2'-substituted nucleoside
  • Inhibitor of HCV replication with antiviral activity
  • Inhibits RNA-dependent RNA polymerase (NS5B)-catalyzed RNA synthesis in vitro
Catalog No. 50-004-52584 Supplier Medchemexpress LLC Supplier No. HYW0118341G
2'-O-Methylcytidine is an orally active 2'-substituted nucleoside that acts as an inhibitor of HCV replication with antiviral activity. It inhibits RNA-dependent RNA polymerase (NS5B)-catalyzed RNA synthesis in vitro by competing with substrate nucleoside triphosphate.
  • Orally active 2'-substituted nucleoside
  • Inhibitor of HCV replication with antiviral activity
  • Inhibits RNA-dependent RNA polymerase (NS5B)-catalyzed RNA synthesis in vitro