Learn More
Medchemexpress LLC 2'-O-Methylcytidine | 2140-72-9 | MFCD00030635 | 99.7% | 257.25 | 500 MG
SDP
Catalog No. 5000453818
Encompass_Preferred
2'-O-Methylcytidine is an orally active 2'-substituted nucleoside that acts as an inhibitor of HCV replication with antiviral activity. It inhibits RNA-dependent RNA polymerase (NS5B)-catalyzed RNA synthesis in vitro, competitively with substrate nucleoside triphosphate. The product is for research use only and not sold to patients.
  • Orally active 2'-substituted nucleoside.
  • HCV replication inhibitor.
  • Possesses antiviral activity.
  • Inhibits RNA-dependent RNA polymerase (NS5B)-catalyzed RNA synthesis in vitro.
  • Competitive inhibitor with substrate nucleoside triphosphate.
Catalog No. 50-004-53818 Supplier Medchemexpress LLC Supplier No. HYW011834500MG
2'-O-Methylcytidine is an orally active 2'-substituted nucleoside that acts as an inhibitor of HCV replication with antiviral activity. It inhibits RNA-dependent RNA polymerase (NS5B)-catalyzed RNA synthesis in vitro, competitively with substrate nucleoside triphosphate. The product is for research use only and not sold to patients.
  • Orally active 2'-substituted nucleoside.
  • HCV replication inhibitor.
  • Possesses antiviral activity.
  • Inhibits RNA-dependent RNA polymerase (NS5B)-catalyzed RNA synthesis in vitro.
  • Competitive inhibitor with substrate nucleoside triphosphate.