Learn More
Apexbio Technology LLC 2'-O-Methylguanosine 2140-71-8 250mg
SDP
Catalog No. 5000093345
Encompass_Preferred
2 -O-Methylguanosine (CAS 2140-71-8) is a modified nucleoside generated enzymatically by tRNA guanosine-2 -O-methyltransferase This compound exhibits biological activity by modulating mRNA translation efficiency and interfering with cell cycle regulation through specific signaling pathways In in vitro studies 2 -O-Methylguanosine has been shown to induce apoptosis making it a valuable molecular tool in cancer research It is frequently utilized to investigate apoptosis induction mechanisms and to support the development of novel therapeutic approaches for related diseases
Catalog No. 50-000-93345 Supplier Apexbio Technology LLC Supplier No. A4769250
2 -O-Methylguanosine (CAS 2140-71-8) is a modified nucleoside generated enzymatically by tRNA guanosine-2 -O-methyltransferase This compound exhibits biological activity by modulating mRNA translation efficiency and interfering with cell cycle regulation through specific signaling pathways In in vitro studies 2 -O-Methylguanosine has been shown to induce apoptosis making it a valuable molecular tool in cancer research It is frequently utilized to investigate apoptosis induction mechanisms and to support the development of novel therapeutic approaches for related diseases