Learn More
Medchemexpress LLC 2'-O-Methylguanosine | 2140-71-8 | 99.8% | 297.27 | 100 MG
SDP
Catalog No. 5000454149
Encompass_Preferred
2'-O-Methylguanosine is a modified nucleoside produced in tRNAs by the action of tRNA guanosine-2'-O-methyltransferase. It results in apoptotic changes of cells and can induce apoptosis in Molt4 cells. It also inhibits the activity of RNA-dependent RNA polymerase after the incorporation of nucleotides, thereby protecting coronavirus RNA from degradation by host nucleases.
  • Modified nucleoside.
  • Apoptosis inducing nucleoside (AIN).
  • Can be isolated from the culture supernatant of CD57⁺HLA-DRbright natural suppressor cell (57.DR-NS).
  • Induces DNA fragmentation and apoptosis in Molt4 cells.
  • Inhibits RNA-dependent RNA polymerase activity.
  • Protects coronavirus RNA from degradation.
Catalog No. 50-004-54149 Supplier Medchemexpress LLC Supplier No. HYW013260100MG
2'-O-Methylguanosine is a modified nucleoside produced in tRNAs by the action of tRNA guanosine-2'-O-methyltransferase. It results in apoptotic changes of cells and can induce apoptosis in Molt4 cells. It also inhibits the activity of RNA-dependent RNA polymerase after the incorporation of nucleotides, thereby protecting coronavirus RNA from degradation by host nucleases.
  • Modified nucleoside.
  • Apoptosis inducing nucleoside (AIN).
  • Can be isolated from the culture supernatant of CD57⁺HLA-DRbright natural suppressor cell (57.DR-NS).
  • Induces DNA fragmentation and apoptosis in Molt4 cells.
  • Inhibits RNA-dependent RNA polymerase activity.
  • Protects coronavirus RNA from degradation.