Learn More
Medchemexpress LLC 2'-O-Methylguanosine | 2140-71-8 | 99.8% | 297.27 | 250 MG
SDP
Catalog No. 5000451409
Encompass_Preferred
2'-O-Methylguanosine is a modified nucleoside produced in tRNAs by the action of tRNA guanosine-2'-O-methyltransferase. It results in apoptotic changes of cells. For research use only; not sold to patients.
  • Modified nucleoside produced in tRNAs
  • Induces apoptotic changes in cells
  • Can be isolated from the culture supernatant of CD57⁺HLA-DRbright natural suppressor cell (57.DR-NS)
  • Induces apoptosis in Molt4 cells at 7 μg/mL for 72 h
  • Inhibits the activity of RNA-dependent RNA polymerase after nucleotide incorporation by hindering translocation
  • Protects coronavirus RNA from degradation by host nucleases
Catalog No. 50-004-51409 Supplier Medchemexpress LLC Supplier No. 250MGHYW013260
2'-O-Methylguanosine is a modified nucleoside produced in tRNAs by the action of tRNA guanosine-2'-O-methyltransferase. It results in apoptotic changes of cells. For research use only; not sold to patients.
  • Modified nucleoside produced in tRNAs
  • Induces apoptotic changes in cells
  • Can be isolated from the culture supernatant of CD57⁺HLA-DRbright natural suppressor cell (57.DR-NS)
  • Induces apoptosis in Molt4 cells at 7 μg/mL for 72 h
  • Inhibits the activity of RNA-dependent RNA polymerase after nucleotide incorporation by hindering translocation
  • Protects coronavirus RNA from degradation by host nucleases