Learn More
Cayman Chemical 2'-O-Methylguanosine, Purity: ≥98%, CAS: 2140-71-8, 250 mg
SDP
Catalog No. 502984343
Encompass_Preferred
A modified nucleoside that is produced in tRNAs by the action of tRNA guanosine-2’-O-methyltransferase
Catalog No. 50-298-4343 Supplier Cayman Chemical Supplier No. 21039250 MG
A modified nucleoside that is produced in tRNAs by the action of tRNA guanosine-2’-O-methyltransferase