Learn More
Medchemexpress LLC 2'-O-Methylisoliquiritigenin | 51828-10-5 | 270.28 | 5 MG
SDP
Catalog No. 5000416014
Encompass_Preferred
2'-O-Methylisoliquiritigenin, isolated from the Arachis species, up-regulates 5-HT, NE, DA, and GABA pathways, though its effect on the NE pathway is not significant. This compound has shown sedative-hypnotic and anxiolytic effects in mice studies and acts on 5-HT, Dopamine, and GABA Receptors, engaging GPCR/G Protein and Neuronal Signaling pathways. It is intended for research use only.
  • Isolated from Arachis species.
  • Up-regulates 5-HT, NE, DA, and GABA pathways.
  • Acts on 5-HT Receptor, Dopamine Receptor, and GABA Receptor.
  • Engages GPCR/G Protein and Neuronal Signaling pathways.
  • Has sedative-hypnotic and anxiolytic effects.
  • Appears as a solid with a light yellow to yellow color.
Catalog No. 50-004-16014 Supplier Medchemexpress LLC Supplier No. HYN17455MG
2'-O-Methylisoliquiritigenin, isolated from the Arachis species, up-regulates 5-HT, NE, DA, and GABA pathways, though its effect on the NE pathway is not significant. This compound has shown sedative-hypnotic and anxiolytic effects in mice studies and acts on 5-HT, Dopamine, and GABA Receptors, engaging GPCR/G Protein and Neuronal Signaling pathways. It is intended for research use only.
  • Isolated from Arachis species.
  • Up-regulates 5-HT, NE, DA, and GABA pathways.
  • Acts on 5-HT Receptor, Dopamine Receptor, and GABA Receptor.
  • Engages GPCR/G Protein and Neuronal Signaling pathways.
  • Has sedative-hypnotic and anxiolytic effects.
  • Appears as a solid with a light yellow to yellow color.