Learn More
Aldevron 2'O-Methyltransferase, 3400ug
Catalog No. 503073877
Encompass_Preferred
3400ug dispense of 2'O-Methyltransferase. 2'-O-Methyltransferase adds a methyl group to the 2'-OH of the first nucleotide adjacent to the m7Gppp5'N cap. 2'-O-Methyltransferase utilizes S-adenosylmethionine as the methyl group donor.
Catalog No. 50-307-3877 Supplier Aldevron Supplier No. 91303400MCG
3400ug dispense of 2'O-Methyltransferase. 2'-O-Methyltransferase adds a methyl group to the 2'-OH of the first nucleotide adjacent to the m7Gppp5'N cap. 2'-O-Methyltransferase utilizes S-adenosylmethionine as the methyl group donor.