Learn More
Aldevron 2'O-Methyltransferase, 1450ug
Catalog No. 503073899
Encompass_Preferred
1450ug dispense of 2'O-Methyltransferase. 2'-O-Methyltransferase adds a methyl group to the 2'-OH of the first nucleotide adjacent to the m7Gppp5'N cap. 2'-O-Methyltransferase utilizes S-adenosylmethionine as the methyl group donor.
Catalog No. 50-307-3899 Supplier Aldevron Supplier No. 91301450MCG
1450ug dispense of 2'O-Methyltransferase. 2'-O-Methyltransferase adds a methyl group to the 2'-OH of the first nucleotide adjacent to the m7Gppp5'N cap. 2'-O-Methyltransferase utilizes S-adenosylmethionine as the methyl group donor.