Learn More
Medchemexpress LLC 2'-O-Methyluridine | 2140-76-3 | 99.8% | 258.23 | 5 G
SDP
Catalog No. 5000367074
Encompass_Preferred
2'-O-Methyluridine is a modified nucleoside that can be found in *T. thermophile* tRNA. Its level in serum is decreased in patients with breast cancer. It can be incorporated into siRNA targeting the gene encoding apolipoprotein B, which decreases ApoB protein levels in HepG2 and prevents IFN-α production in isolated human PBMCs.
  • Found in *T. thermophile* tRNA
  • Decreased serum levels in breast cancer patients
  • Can be incorporated into siRNA
  • Decreases ApoB protein levels in HepG2
  • Prevents IFN-α production in isolated human PBMCs
Catalog No. 50-003-67074 Supplier Medchemexpress LLC Supplier No. HYW0118245G
2'-O-Methyluridine is a modified nucleoside that can be found in *T. thermophile* tRNA. Its level in serum is decreased in patients with breast cancer. It can be incorporated into siRNA targeting the gene encoding apolipoprotein B, which decreases ApoB protein levels in HepG2 and prevents IFN-α production in isolated human PBMCs.
  • Found in *T. thermophile* tRNA
  • Decreased serum levels in breast cancer patients
  • Can be incorporated into siRNA
  • Decreases ApoB protein levels in HepG2
  • Prevents IFN-α production in isolated human PBMCs