Learn More
Medchemexpress LLC 2'-O-Methyluridine-d3 | 2140-76-3 | 99.2% | 261.25 | 1 MG
SDP
Catalog No. 5000453817
Encompass_Preferred
2'-O-Methyluridine-d3 is deuterium labeled 2'-O-Methyluridine (HY-W011824). 2'-O-Methyluridine is a modified nucleoside found in *T. thermophile* tRNA. It can be incorporated into siRNA targeting the gene encoding apolipoprotein B, which decreases ApoB protein levels in HepG2 and prevents IFN-α production in isolated human PBMCs. The level of 2'-O-Methyluridine in serum is decreased in patients with breast cancer. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-53817 Supplier Medchemexpress LLC Supplier No. HYW011824S1MG
2'-O-Methyluridine-d3 is deuterium labeled 2'-O-Methyluridine (HY-W011824). 2'-O-Methyluridine is a modified nucleoside found in *T. thermophile* tRNA. It can be incorporated into siRNA targeting the gene encoding apolipoprotein B, which decreases ApoB protein levels in HepG2 and prevents IFN-α production in isolated human PBMCs. The level of 2'-O-Methyluridine in serum is decreased in patients with breast cancer. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS