Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-O-MOE-5-Me-rC | 244105-55-3 | 98.0% | 315.32 g/mol | C13H21N3O6 | 250 MG
SDP

Catalog No. 5000209409
Encompass_Preferred

2'-O-MOE-5-Me-rC is a 2'-O-methoxyethyl and 5-methyl modified ribonucleoside used for incorporation into synthetic oligonucleotides to improve hybridization and nuclease resistance.

  • Modified ribonucleoside suitable for oligonucleotide synthesis.
  • 2'-O-methoxyethyl (MOE) and 5-methyl modifications enhance binding affinity.
  • High purity: 98.0%.
  • Available in multiple sizes, e.g. 100 mg, 250 mg, 500 mg, 1 g.
  • Molecular formula C13H21N3O6; molecular weight 315.32 g/mol.
  • Store solid at 4°C and protect from light; in solvent store at -80°C up to 6 months.

Catalog No. 50-002-09409 Supplier Medchemexpress LLC Supplier No. 250MGHYW048497
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-O-MOE-5-Me-rC is a 2'-O-methoxyethyl and 5-methyl modified ribonucleoside used for incorporation into synthetic oligonucleotides to improve hybridization and nuclease resistance.

  • Modified ribonucleoside suitable for oligonucleotide synthesis.
  • 2'-O-methoxyethyl (MOE) and 5-methyl modifications enhance binding affinity.
  • High purity: 98.0%.
  • Available in multiple sizes, e.g. 100 mg, 250 mg, 500 mg, 1 g.
  • Molecular formula C13H21N3O6; molecular weight 315.32 g/mol.
  • Store solid at 4°C and protect from light; in solvent store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.