Learn More
Medchemexpress LLC 2'-O-MOE-5MeU-3'-phosphoramidite | 163878-63-5 | MFCD11505865 | ≥97.0% | 818.89 | C43H55N4O10P | 250 MG
SDP
Catalog No. 5000241849
Encompass_Preferred
2'-O-MOE-5MeU-3'-phosphoramidite is a phosphoramidite monomer used to incorporate 2'-O-methoxyethyl-modified 5-methyluridine into oligonucleotides by automated solid-phase synthesis. It is supplied as a white to off-white solid (molecular weight 818.89; formula C43H55N4O10P) with a typical purity of ≥97% and is offered in multiple pack sizes for research use.
  • Compatible with automated solid-phase oligonucleotide synthesis.
  • Enables incorporation of 2'-O-methoxyethyl-modified 5-methyluridine residues.
  • Supplied as a white to off-white solid with documented molecular weight and formula.
  • Typical purity ≥97% suitable for research applications.
  • Available in multiple pack sizes to support different synthesis scales.
Catalog No. 50-002-41849 Supplier Medchemexpress LLC Supplier No. HY104016250MG
2'-O-MOE-5MeU-3'-phosphoramidite is a phosphoramidite monomer used to incorporate 2'-O-methoxyethyl-modified 5-methyluridine into oligonucleotides by automated solid-phase synthesis. It is supplied as a white to off-white solid (molecular weight 818.89; formula C43H55N4O10P) with a typical purity of ≥97% and is offered in multiple pack sizes for research use.
  • Compatible with automated solid-phase oligonucleotide synthesis.
  • Enables incorporation of 2'-O-methoxyethyl-modified 5-methyluridine residues.
  • Supplied as a white to off-white solid with documented molecular weight and formula.
  • Typical purity ≥97% suitable for research applications.
  • Available in multiple pack sizes to support different synthesis scales.