Learn More
Medchemexpress LLC 2'-O-MOE-rC | 251647-54-8 | MFCD15145377 | 98.0% | 907.99 g/mol | C49H58N5O10P | 250 MG
SDP
Catalog No. 5000284126
Encompass_Preferred
2'-O-MOE-rC is a 2'-O-methoxyethyl modified cytidine nucleoside used as a building block for DNA and RNA oligonucleotide synthesis. It is intended for research use and is supplied as a solid powder suitable for incorporation into custom oligonucleotides, offering enhanced stability and performance compared with unmodified nucleosides.
  • Used as a building block for oligonucleotide synthesis.
  • Confers increased nuclease resistance compared with unmodified nucleosides.
  • Improves hybridization affinity and binding specificity of modified oligonucleotides.
  • Supplied as a solid powder, commonly available in 250 MG packages.
  • Stable when stored at 4°C; in solvent, store at -80°C for long-term storage.
Catalog No. 50-002-84126 Supplier Medchemexpress LLC Supplier No. HYW570885250MG
2'-O-MOE-rC is a 2'-O-methoxyethyl modified cytidine nucleoside used as a building block for DNA and RNA oligonucleotide synthesis. It is intended for research use and is supplied as a solid powder suitable for incorporation into custom oligonucleotides, offering enhanced stability and performance compared with unmodified nucleosides.
  • Used as a building block for oligonucleotide synthesis.
  • Confers increased nuclease resistance compared with unmodified nucleosides.
  • Improves hybridization affinity and binding specificity of modified oligonucleotides.
  • Supplied as a solid powder, commonly available in 250 MG packages.
  • Stable when stored at 4°C; in solvent, store at -80°C for long-term storage.