Learn More
Cayman Chemical 2-O-MonsuccInyladenosIn-3 10mg
SDP
Catalog No. 502992396
Encompass_Preferred
An immunogenic derivative of cAMP; has been used to generate anti-cAMP antisera and antibodies
Catalog No. 50-299-2396 Supplier Cayman Chemical Supplier No. 3053510 MG
An immunogenic derivative of cAMP; has been used to generate anti-cAMP antisera and antibodies