Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 2'-O-NMA MeC(Bz) amidite | 1 g | CAS 1025783-18-9 | InChIKey OABRWFIIVWCDDM-WBHVIXSUSA-N

Catalog No. 502364258
Encompass

2'-O-NMA MeC(Bz) amidite is a Amidite reagent (Subcategory: 2'-NMA Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 1025783-18-9
- MDL: No data
- InChIKey: OABRWFIIVWCDDM-WBHVIXSUSA-N
- Molecular Weight: 935.027761998
- Molecular Formula: C50H59N6O10P
- Purity: ≥95%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-5-(4-benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(2-(methylamino)-2-oxoethoxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]([C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C=C(C(NC(C6=CC=CC=C6)=O)=NC5=O)C)OCC(NC)=O)OCCC#N)C(C)C)C

Catalog No. 50-236-4258 Supplier STA Pharmaceutical Supplier No. STAAM10692AK601GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-O-NMA MeC(Bz) amidite is a Amidite reagent (Subcategory: 2'-NMA Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 1025783-18-9
- MDL: No data
- InChIKey: OABRWFIIVWCDDM-WBHVIXSUSA-N
- Molecular Weight: 935.027761998
- Molecular Formula: C50H59N6O10P
- Purity: ≥95%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-5-(4-benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(2-(methylamino)-2-oxoethoxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]([C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C=C(C(NC(C6=CC=CC=C6)=O)=NC5=O)C)OCC(NC)=O)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.