Learn More
Cayman Chemical 2''-O-Rhamnosylicariside II, Purity: ≥98%, CAS: 135293-13-9, 10 mg
SDP
Catalog No. 502912064
Encompass_Preferred
A flavonoid glycoside with hepatotoxic activity; increases AST, LDH, and MDA levels in Hl 7702 cells at 66 µg/ml; induces 100% mortality in zebrafish larvae at 100 µM; induces hepatocellular vacuolization and degeneration in adult zebrafish at 25 µM
Catalog No. 50-291-2064 Supplier Cayman Chemical Supplier No. 3648710 MG
A flavonoid glycoside with hepatotoxic activity; increases AST, LDH, and MDA levels in Hl 7702 cells at 66 µg/ml; induces 100% mortality in zebrafish larvae at 100 µM; induces hepatocellular vacuolization and degeneration in adult zebrafish at 25 µM