Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+/-)-2-Octanol

Catalog No. 7485825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-858-25ML 25 mL
74-858-100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-858-25ML Supplier Sigma Aldrich Supplier No. 7485825ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reagentplus is a registered trademark of sigma-aldrich co
  • Llc

Specifications

CAS 123-96-6
Density 0.819 g/mL (at 20°C (literature))
Molecular Formula C8H18O
Refractive Index n20/D 1.426
Linear Formula CH3(CH2)5 CH(OH)CH3
MDL Number MFCD00004591
Quantity 25 mL
Synonym Hexyl methyl carbinol
Molecular Weight (g/mol) 130.23
Percent Purity ≥99.5% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.