Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-2-Octanol, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019691 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9691 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9691 Supplier MilliporeSigma Supelco Supplier No. 422771ML
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 123-96-6
Density 0.819 g/mL (at 20°C)
Flash Point 71°C
Physical Form Neat
Linear Formula CH3(CH2)5 CH(OH)CH3
MDL Number MFCD00004591
Synonym Hexyl methyl carbinol
Type Analytical Standard
Quantity 1 mL
Formula Weight 130.23
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.