Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-Octanol, 98%

Catalog No. AAA169850F
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA169850F 2500 mL
AAA16985AE 100 mL
AAA16985AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA169850F Supplier Thermo Scientific Chemicals Supplier No. A169850F
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 123-96-6 | C8H18O | 130.231 g/mol

(±)-2-Octanol is used in flavors, paints and coatings, defoaming agent, low-volatility solvent, lubricants, fuels, paints and coatings. It is used as a frother in mineral floatation and wetting agent. It acts as an intermediate in the preparation of surfactants, cosmetic emollients esters and plasticizers. Further, it is also used as a reagent in the synthesis of pharmaceutical compounds and piperine derivatives. In addition to this, it is considered as an alternative of 2-ethylhexanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-2-Octanol is used in flavors, paints and coatings, defoaming agent, low-volatility solvent, lubricants, fuels, paints and coatings. It is used as a frother in mineral floatation and wetting agent. It acts as an intermediate in the preparation of surfactants, cosmetic emollients esters and plasticizers. Further, it is also used as a reagent in the synthesis of pharmaceutical compounds and piperine derivatives. In addition to this, it is considered as an alternative of 2-ethylhexanol.

Solubility
Miscible with diethyl ether, acetone, alcohol, most common organic solvents, aromatic and aliphatic hydrocarbons. Slightly miscible with cold water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-96-6
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD00004591
InChI Key SJWFXCIHNDVPSH-UHFFFAOYSA-N
Synonym 2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
PubChem CID 20083
ChEBI CHEBI:37869
IUPAC Name octan-2-ol
SMILES CCCCCCC(C)O

Specifications

Melting Point -38°C
Density 0.819
Boiling Point 179°C to 180°C
Flash Point 76°C (169°F)
Odor Pleasant
Refractive Index 1.426
Quantity 2500 mL
Beilstein 1719325
Merck Index 14,6752
Solubility Information Miscible with diethyl ether,acetone,alcohol,most common organic solvents,aromatic and aliphatic hydrocarbons. Slightly miscible with cold water.
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material (+/-)-2-Octanol
Show More Show Less
Hazard Category H227-H318
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P280-P305+P351+P338-P310-P370+P378q-P501c
EINECSNumber 204-667-0
RTECSNumber RH0795000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.