Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Octanone

Catalog No. O470925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O470925G 25 g
O47092KG 2 kg
O4709500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O470925G Supplier Sigma Aldrich Supplier No. O470925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Octanone

Specifications

CAS 111-13-7
Melting Point -16°C (lit.)
Density 0.819 g/mL (at 25°C (literature))
Boiling Point 173°C (lit.)
Molecular Formula C8H16O
Refractive Index n20/D 1.416 (literature)
Linear Formula CH3(CH2)5 COCH3
MDL Number MFCD00009540
Quantity 25 g
Synonym Hexyl methyl ketone
Molecular Weight (g/mol) 128.21
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.