Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-octen-1-ylsuccinic anhydride, mixture of cis and trans | 42482-06-4 | MFCD00047133 | >=97.0% | 210.27 | C12H18O3 | 100 G
SDP

Catalog No. 5000283647
Encompass_Preferred

2-Octen-1-ylsuccinic anhydride (mixture of cis and trans) is an alkenyl succinic anhydride reagent used for acylation and functionalization in organic synthesis. It is commonly employed to introduce succinic anhydride moieties into polysaccharides and small molecules to produce amphiphilic derivatives for materials and biochemical research. The material is supplied as a colorless to light yellow liquid with good solubility in DMSO.

  • Mixture of cis and trans isomers
  • CAS number 42482-06-4
  • Molecular formula C12H18O3; molecular weight 210.27
  • Appearance: colorless to light yellow liquid
  • Soluble in DMSO at ≥200 mg/mL
  • Storage stability: room temperature for up to 3 years
  • Common pack sizes: 25 G, 50 G, 100 G

Catalog No. 50-002-83647 Supplier Medchemexpress LLC Supplier No. HYW128533100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Octen-1-ylsuccinic anhydride (mixture of cis and trans) is an alkenyl succinic anhydride reagent used for acylation and functionalization in organic synthesis. It is commonly employed to introduce succinic anhydride moieties into polysaccharides and small molecules to produce amphiphilic derivatives for materials and biochemical research. The material is supplied as a colorless to light yellow liquid with good solubility in DMSO.

  • Mixture of cis and trans isomers
  • CAS number 42482-06-4
  • Molecular formula C12H18O3; molecular weight 210.27
  • Appearance: colorless to light yellow liquid
  • Soluble in DMSO at ≥200 mg/mL
  • Storage stability: room temperature for up to 3 years
  • Common pack sizes: 25 G, 50 G, 100 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.