Learn More
Medchemexpress LLC 2-Octen-1-ylsuccinic anhydride, mixture of cis and trans | 42482-06-4 | MFCD00047133 | ≥97.0% | 210.27 g·mol⁻¹ | C12H18O3 | 25 G
SDP
Supplier:  Medchemexpress LLC HYW12853325G
Encompass_Preferred
2-Octen-1-ylsuccinic anhydride (mixture of cis and trans) is an anhydride reagent used to introduce octenyl succinyl groups onto polysaccharides and other hydroxyl-containing substrates. It is commonly used to prepare amphiphilic pectin oligosaccharide derivatives and to modify starches or polymers in laboratory research.
  • Used to prepare amphiphilic pectin oligosaccharide derivatives.
  • Introduces octenyl succinyl groups onto hydroxyl-containing substrates.
  • Mixture of cis and trans isomers for broader reactivity.
  • High purity (≥97.0%).
  • Available in small pack sizes suitable for lab-scale work.
  • Store as recommended to maintain stability.
Catalog No. 50-002-82753