Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-octen-1-ylsuccinic anhydride, mixture of cis and trans | 42482-06-4 | MFCD00047133 | 97.0% | 210.27 g/mol | C12H18O3 | 50 G
SDP

Catalog No. 5000283434
Encompass_Preferred

2-Octen-1-ylsuccinic anhydride (mixture of cis and trans) is a liquid anhydride reagent (C12H18O3, 210.27 g/mol; CAS 42482-06-4) used in research to derivatize polysaccharides and to prepare amphiphilic pectin oligosaccharide derivatives. The material is supplied with manufacturer documentation and is offered in multiple pack sizes for laboratory use.

  • Liquid, colorless to light yellow.
  • Molecular formula C12H18O3; molecular weight 210.27 g/mol.
  • Used to prepare amphiphilic pectin oligosaccharide derivatives.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
  • Storage: store neat at room temperature; in solvent keep at -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-83434 Supplier Medchemexpress LLC Supplier No. HYW12853350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Octen-1-ylsuccinic anhydride (mixture of cis and trans) is a liquid anhydride reagent (C12H18O3, 210.27 g/mol; CAS 42482-06-4) used in research to derivatize polysaccharides and to prepare amphiphilic pectin oligosaccharide derivatives. The material is supplied with manufacturer documentation and is offered in multiple pack sizes for laboratory use.

  • Liquid, colorless to light yellow.
  • Molecular formula C12H18O3; molecular weight 210.27 g/mol.
  • Used to prepare amphiphilic pectin oligosaccharide derivatives.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
  • Storage: store neat at room temperature; in solvent keep at -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.