Learn More
Medchemexpress LLC 2-Octyl-4(1H)-quinolone | 80554-60-5 | ≥98.0% | 257.37 | 100 MG
SDP
Catalog No. 5000404671
Encompass_Preferred
2-Octyl-4(1H)-quinolone, also known as Compound 14, is a quinolinone alkaloid that can be isolated from the plant Ruta graveolens. It is intended for research use only.
  • It is a natural product derived from Ruta graveolens.
  • It exhibits antiproliferative activity against human A2780 cells.
  • The IC50 value for its antiproliferative activity against A2780 cells is 12 μM after 2 days by Alamar blue assay.
Catalog No. 50-004-04671 Supplier Medchemexpress LLC Supplier No. HY402321100MG
2-Octyl-4(1H)-quinolone, also known as Compound 14, is a quinolinone alkaloid that can be isolated from the plant Ruta graveolens. It is intended for research use only.
  • It is a natural product derived from Ruta graveolens.
  • It exhibits antiproliferative activity against human A2780 cells.
  • The IC50 value for its antiproliferative activity against A2780 cells is 12 μM after 2 days by Alamar blue assay.