Learn More
Medchemexpress LLC 2-Octyl-4(1H)-quinolone | 80554-60-5 | ≥98.0% | 257.37 | 25 MG
SDP
Catalog No. 5000402311
Encompass_Preferred
2-Octyl-4(1H)-quinolone (Compound 14) is a quinolinone alkaloid that can be isolated from *Ruta graveolens*. It has antiproliferative activity against human A2780 cells with an IC50 of 12 μM.
  • Quinolinone alkaloid
  • Isolated from *Ruta graveolens*
  • Antiproliferative activity against human A2780 cells
Catalog No. 50-004-02311 Supplier Medchemexpress LLC Supplier No. 25MGHY402321
2-Octyl-4(1H)-quinolone (Compound 14) is a quinolinone alkaloid that can be isolated from *Ruta graveolens*. It has antiproliferative activity against human A2780 cells with an IC50 of 12 μM.
  • Quinolinone alkaloid
  • Isolated from *Ruta graveolens*
  • Antiproliferative activity against human A2780 cells