Learn More
Medchemexpress LLC 2-Octyl-4(1H)-quinolone | 80554-60-5 | >=98.0% | 257.37 | 50 MG
SDP
Catalog No. 5000401618
Encompass_Preferred
2-Octyl-4(1H)-quinolone is a quinolinone alkaloid that can be isolated from *Ruta graveolens*. It is intended for research use only. It has shown antiproliferative activity against human A2780 cells with an IC50 of 12 μM after 2 days by Alamar blue assay. The compound is a solid, white to off-white in color, with the formula C17H23NO. It is classified under Alkaloids and Other Alkaloids, and its initial source is from the plants Rutaceae, specifically *Ruta graveolens Linn.*.
  • Quinolone alkaloid.
  • Isolated from *Ruta graveolens*.
  • For research use only.
  • Exhibits antiproliferative activity against human A2780 cells with an IC50 of 12 μM after 2 days by Alamar blue assay.
  • Solid, white to off-white in color.
  • Chemical formula C17H23NO.
  • Classified under alkaloids.
  • Sourced from Rutaceae, *Ruta graveolens Linn.*.
Catalog No. 50-004-01618 Supplier Medchemexpress LLC Supplier No. HY40232150MG
2-Octyl-4(1H)-quinolone is a quinolinone alkaloid that can be isolated from *Ruta graveolens*. It is intended for research use only. It has shown antiproliferative activity against human A2780 cells with an IC50 of 12 μM after 2 days by Alamar blue assay. The compound is a solid, white to off-white in color, with the formula C17H23NO. It is classified under Alkaloids and Other Alkaloids, and its initial source is from the plants Rutaceae, specifically *Ruta graveolens Linn.*.
  • Quinolone alkaloid.
  • Isolated from *Ruta graveolens*.
  • For research use only.
  • Exhibits antiproliferative activity against human A2780 cells with an IC50 of 12 μM after 2 days by Alamar blue assay.
  • Solid, white to off-white in color.
  • Chemical formula C17H23NO.
  • Classified under alkaloids.
  • Sourced from Rutaceae, *Ruta graveolens Linn.*.