Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octylamine, 98%, Thermo Scientific™

Catalog No. AC179950250
Change view
Click to view available options
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC179950250 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC179950250 Supplier Thermo Scientific Chemicals Supplier No. 179950250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 693-16-3
CAS Min % 97.5
CAS Max % 100.0
Density 0.771g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C8H19N
Refractive Index 1.4225 to 1.4245 (20°C, 589nm)
Linear Formula CH3(CH2)5CH(CH3)NH2
Quantity 25mL
Beilstein 04, 196
Synonym 2-aminooctane, 2-octanamine, 2-octylamine, 1-methylheptylamine, heptylamine, 1-methyl, 2-caprylamine, sec-octylamine, 1-methyl heptylamine, oct-2-ylamine, sec-octanamine
InChI Key HBXNJMZWGSCKPW-UHFFFAOYSA-N
SMILES CCCCCCC(C)N
IUPAC Name octan-2-amine
Molecular Weight (g/mol) 129.247
PubChem CID 12735
Formula Weight 129.24
Percent Purity ≥97.5%
Chemical Name or Material 2-Octylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-744-2
RTECSNumber MK1210000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.