Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octyne 99.0+%, TCI America™
SDP

Catalog No. O01255ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01255ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01255ML Supplier TCI America Supplier No. O01255ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2809-67-8
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00027302
InChI Key QCQALVMFTWRCFI-UHFFFAOYSA-N
Synonym 2-octyne, n-c5h11c.$.cch3, amylmethylacetylene, methylpentylacetylene, acmc-209h0u, n-c5h11c.equiv.cch3
PubChem CID 17769
IUPAC Name oct-2-yne
SMILES CCCCCC#CC

Specifications

Color Colorless
Boiling Point 137°C
Quantity 5 mL
UN Number 3295
Formula Weight 110.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Octyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.