Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octynoic acid, 98%

Catalog No. aab2195722
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2195722 100 g
AAB2195706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2195722 Supplier Thermo Scientific Chemicals Supplier No. B2195722
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5663-96-7 | C8H12O2 | 140.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5663-96-7
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00004364
InChI Key BQDKCWCMDBMLEH-UHFFFAOYSA-N
Synonym 2-octynoic acid, 2-octyn-1-oic acid, m-pentyl propiolate, propiolic acid, pentyl, propiolic acid, pentyl ester, pentylpropiolic acid, 2-oa, 2-octynoicacid, acmc-1apg2, epitope id:137475
PubChem CID 21872
ChEBI CHEBI:60295
IUPAC Name oct-2-ynoic acid
SMILES CCCCCC#CC(=O)O

Specifications

Density 0.961
Boiling Point 148°C to 149°C (19 mmHg)
Flash Point 101°C (214°F)
Refractive Index 1.46
Quantity 100 g
UN Number UN3265
Formula Weight 140.18
Percent Purity 98%
Chemical Name or Material 2-Octynoic acid
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 227-121-3
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.