Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Octynoic acid, methyl ester | 111-12-6 | 99.8% | 154.21 | 10 G
SDP

Catalog No. 5000454729
Encompass_Preferred

Methyl 2-Octynoate is a hapten intended for research use only, not for sale to patients.

  • Appearance: Liquid, colorless to light yellow
  • Formula: C9H14O2
  • Molecular weight: 154.21
  • Purity: 99.78%
  • CAS number: 111-12-6
  • Storage: Pure form at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-54729 Supplier Medchemexpress LLC Supplier No. HYW01357610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 2-Octynoate is a hapten intended for research use only, not for sale to patients.

  • Appearance: Liquid, colorless to light yellow
  • Formula: C9H14O2
  • Molecular weight: 154.21
  • Purity: 99.78%
  • CAS number: 111-12-6
  • Storage: Pure form at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.