Learn More
Cayman Chemical 2-Oleoyl Glycerol 10mg
SDP
Catalog No. 502980277
Encompass_Preferred
A natural monoacylglycerol that is metabolized by monoacylglycerol lipase; an agonist of GPR119 (EC50 = 2.5 µM), stimulating the release of glucagon-like peptide-1 from intestinal L-cells
Catalog No. 50-298-0277 Supplier Cayman Chemical Supplier No. 1653710 MG
A natural monoacylglycerol that is metabolized by monoacylglycerol lipase; an agonist of GPR119 (EC50 = 2.5 µM), stimulating the release of glucagon-like peptide-1 from intestinal L-cells