Learn More
Cayman Chemical 2-Oleoyl Glycerol, Purity: ≥98%, CAS: 3443-84-3, 1 mg
SDP
Catalog No. 502980636
Encompass_Preferred
A natural monoacylglycerol that is metabolized by monoacylglycerol lipase; an agonist of GPR119 (EC50 = 2.5 µM), stimulating the release of glucagon-like peptide-1 from intestinal L-cells
Catalog No. 50-298-0636 Supplier Cayman Chemical Supplier No. 165371 MG
A natural monoacylglycerol that is metabolized by monoacylglycerol lipase; an agonist of GPR119 (EC50 = 2.5 µM), stimulating the release of glucagon-like peptide-1 from intestinal L-cells