Learn More
Cayman Chemical 2-Oleoyl Glycerol 5mg
SDP
Catalog No. 502980046
Encompass_Preferred
A natural monoacylglycerol that is metabolized by monoacylglycerol lipase; an agonist of GPR119 (EC50 = 2.5 µM), stimulating the release of glucagon-like peptide-1 from intestinal L-cells
Catalog No. 50-298-0046 Supplier Cayman Chemical Supplier No. 165375 MG
A natural monoacylglycerol that is metabolized by monoacylglycerol lipase; an agonist of GPR119 (EC50 = 2.5 µM), stimulating the release of glucagon-like peptide-1 from intestinal L-cells