Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 2'-OMe C(Ac) PACE amidite | 5 g | CAS 1355047-25-4 | InChIKey RQLMSVYEXLXTTN-YQKQXIHOSA-N

Catalog No. 502355180
Encompass

2'-OMe C(Ac) PACE amidite is a Amidite reagent (Subcategory: PACE Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 1355047-25-4
- MDL: No data
- InChIKey: RQLMSVYEXLXTTN-YQKQXIHOSA-N
- Molecular Weight: 871.968761998
- Molecular Formula: C46H58N5O10P
- Purity: ≥98%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 1-cyano-2-methylpropan-2-yl 2-((((2R,3R,4R,5R)-5-(4-acetamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-methoxytetrahydrofuran-3-yl)oxy)(diisopropylamino)phosphaneyl)acetate
- SMILES: O=C(CP(O[C@H]1[C@H]([C@H](N2C(N=C(C=C2)NC(C)=O)=O)O[C@@H]1COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OC)N(C(C)C)C(C)C)OC(C)(CC#N)C

Catalog No. 50-235-5180 Supplier STA Pharmaceutical Supplier No. STAAM100505GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-OMe C(Ac) PACE amidite is a Amidite reagent (Subcategory: PACE Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 1355047-25-4
- MDL: No data
- InChIKey: RQLMSVYEXLXTTN-YQKQXIHOSA-N
- Molecular Weight: 871.968761998
- Molecular Formula: C46H58N5O10P
- Purity: ≥98%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: 1-cyano-2-methylpropan-2-yl 2-((((2R,3R,4R,5R)-5-(4-acetamido-2-oxopyrimidin-1(2H)-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-methoxytetrahydrofuran-3-yl)oxy)(diisopropylamino)phosphaneyl)acetate
- SMILES: O=C(CP(O[C@H]1[C@H]([C@H](N2C(N=C(C=C2)NC(C)=O)=O)O[C@@H]1COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OC)N(C(C)C)C(C)C)OC(C)(CC#N)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.