Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glen Research 2'-O-Methylguanosine CE Phosphoramidite, 1.0 g, C44H55N8O8P, nuclease resistant, optimized for DNA synthesizers

Catalog No. NC3673054
Encompass_Preferred

2'-O-Methylguanosine CE Phosphoramidite is a specialized phosphoramidite monomer designed for the synthesis of oligonucleotides. This reagent enables the incorporation of nuclease-resistant 2'-O-methyl ribonucleotide linkages, ensuring high performance in automated DNA synthesizers. Its deprotection, isolation, and handling procedures are compatible with those used for standard oligodeoxynucleotides.

  • Nuclease resistant linkages
  • Compatible with standard deprotection and handling procedures
  • Optimized packaging for DNA synthesizers
  • Accompanied by certificate of analysis and HPLC trace
  • Molecular formula: C44H55N8O8P
  • Molecular weight: 854.93
  • Application: Oligonucleotide synthesis

Catalog No. NC3673054 Supplier Glen Research Supplier No. 10312110E
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-O-Methylguanosine CE Phosphoramidite is a specialized phosphoramidite monomer designed for the synthesis of oligonucleotides. This reagent enables the incorporation of nuclease-resistant 2'-O-methyl ribonucleotide linkages, ensuring high performance in automated DNA synthesizers. Its deprotection, isolation, and handling procedures are compatible with those used for standard oligodeoxynucleotides.

  • Nuclease resistant linkages
  • Compatible with standard deprotection and handling procedures
  • Optimized packaging for DNA synthesizers
  • Accompanied by certificate of analysis and HPLC trace
  • Molecular formula: C44H55N8O8P
  • Molecular weight: 854.93
  • Application: Oligonucleotide synthesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.