Learn More
Medchemexpress LLC 2'-OMe-G(ibu) Phosphoramidite | 150780-67-9 | 99.4% | 869.94 | 1 G
SDP
Catalog No. 5000349843
Encompass_Preferred
2'-OMe-G(ibu) Phosphoramidite, also known as DMT-2'-O-Methyl-rG(ib) Phosphoramidite, is a modified phosphoramidite monomer critical for oligonucleotide synthesis. It functions effectively as a solid support under standard conditions, suitable for both DNA and 2'-OMe RNA synthesis. This high-purity compound (99.4%) ensures reliable results in various molecular biology applications. It has a molecular weight of 869.94 and a CAS Number of 150780-67-9.
  • Modified phosphoramidite monomer
  • Used for oligonucleotide synthesis
  • Effective as a solid support in DNA and 2'-OMe RNA synthesis
  • High purity for reliable experimental outcomes
  • Optimal storage for solid form is -20°C, protected from light
  • Optimal storage for solution form is -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-003-49843 Supplier Medchemexpress LLC Supplier No. HY1385791G
2'-OMe-G(ibu) Phosphoramidite, also known as DMT-2'-O-Methyl-rG(ib) Phosphoramidite, is a modified phosphoramidite monomer critical for oligonucleotide synthesis. It functions effectively as a solid support under standard conditions, suitable for both DNA and 2'-OMe RNA synthesis. This high-purity compound (99.4%) ensures reliable results in various molecular biology applications. It has a molecular weight of 869.94 and a CAS Number of 150780-67-9.
  • Modified phosphoramidite monomer
  • Used for oligonucleotide synthesis
  • Effective as a solid support in DNA and 2'-OMe RNA synthesis
  • High purity for reliable experimental outcomes
  • Optimal storage for solid form is -20°C, protected from light
  • Optimal storage for solution form is -80°C for 6 months or -20°C for 1 month, protected from light