Learn More
Glen Research 2’-OMe-G-Thiophosphoramidite, 0.25 g
Catalog No. NC1845368
Encompass_Preferred
2’-OMe- RNA Thiophosphoramidites are RNA monomers designed to produce oligos combining the PS2 linkage with the 2’-O-methyl ribose modification. These PS2-modified RNA oligos have potential for use in siRNAs and dithiophosphate aptamers (thioaptamers).
Catalog No. NC1845368 Supplier Glen Research Supplier No. 10317202
2’-OMe- RNA Thiophosphoramidites are RNA monomers designed to produce oligos combining the PS2 linkage with the 2’-O-methyl ribose modification. These PS2-modified RNA oligos have potential for use in siRNAs and dithiophosphate aptamers (thioaptamers).