Learn More
Glen Research 2'-OMe-U-RNA-CPG, 1 μmole per column, 4 columns
Catalog No. NC3725760
Encompass_Preferred
This 2'-O-methyl Uridine RNA CPG support is designed for synthesizing oligonucleotides with enhanced nuclease resistance. It functions similarly to standard nucleoside supports due to its 5'-Dimethoxytrityl (DMT) group, allowing for simplified deprotection, isolation, and handling comparable to those for oligodeoxynucleotides.
  • Nucleoside: Uridine, 2'-O-methyl
  • Support type: Controlled Pore Glass (CPG)
  • Linker: 3'-succinoyl-long chain alkylamino
  • Protecting group: 5'-Dimethoxytrityl (DMT)
  • Application: Oligonucleotide synthesis
  • Feature: Nuclease resistant linkages
  • Synthesis scale: 1 μmole (per column)
  • Pack size: 4 columns
Catalog No. NC3725760 Supplier Glen Research Supplier No. 20373041E
This 2'-O-methyl Uridine RNA CPG support is designed for synthesizing oligonucleotides with enhanced nuclease resistance. It functions similarly to standard nucleoside supports due to its 5'-Dimethoxytrityl (DMT) group, allowing for simplified deprotection, isolation, and handling comparable to those for oligodeoxynucleotides.
  • Nucleoside: Uridine, 2'-O-methyl
  • Support type: Controlled Pore Glass (CPG)
  • Linker: 3'-succinoyl-long chain alkylamino
  • Protecting group: 5'-Dimethoxytrityl (DMT)
  • Application: Oligonucleotide synthesis
  • Feature: Nuclease resistant linkages
  • Synthesis scale: 1 μmole (per column)
  • Pack size: 4 columns