Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 2'-OTBS N1-Me-A(ClAc) amidite | 5 g | InChIKey ISUKAHYBQLYEAJ-VEJAPVAKSA-N | MW 974.608761998

Catalog No. 502364895
Encompass

2'-OTBS N1-Me-A(ClAc) amidite is a Amidite reagent (Subcategory: Adenine Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: ISUKAHYBQLYEAJ-VEJAPVAKSA-N
- Molecular Weight: 974.608761998
- Molecular Formula: C49H65ClN7O8PSi
- Purity: ≥95%
- Container Type: 250 mL Glass (28-400)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 174.7 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-5-(6-((2-chloroacetyl)imino)-1-methyl-1,6-dihydro-9H-purin-9-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]([C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C(N=CN/6C)=C(N=C5)C6=N\C(CCl)=O)O[Si](C)(C(C)(C)C)C)OCCC#N)C(C)C)C

Catalog No. 50-236-4895 Supplier STA Pharmaceutical Supplier No. STAAM10526MM2505GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-OTBS N1-Me-A(ClAc) amidite is a Amidite reagent (Subcategory: Adenine Series) sold by WuXi TIDES. Offered in 5 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: No data
- MDL: No data
- InChIKey: ISUKAHYBQLYEAJ-VEJAPVAKSA-N
- Molecular Weight: 974.608761998
- Molecular Formula: C49H65ClN7O8PSi
- Purity: ≥95%
- Container Type: 250 mL Glass (28-400)
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 174.7 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-5-(6-((2-chloroacetyl)imino)-1-methyl-1,6-dihydro-9H-purin-9-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1[C@H]([C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C(N=CN/6C)=C(N=C5)C6=N\C(CCl)=O)O[Si](C)(C(C)(C)C)C)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.