Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC 2’-OTBS Pyrimidinone amidite | 1 g | CAS 155831-90-6 | MDL MFCD08062082

Catalog No. 502364673
Encompass

2’-OTBS Pyrimidinone amidite is a Amidite reagent (Subcategory: Specialty Base Modification) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 155831-90-6
- MDL: MFCD08062082
- InChIKey: UEUDLVPTCOPGMZ-MGVQLQBBSA-N
- Molecular Weight: 845.061761998
- Molecular Formula: C45H61N4O8PSi
- Purity: ≥95%
- Container Type: 60 mL Glass (20-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 63 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-5-(2-oxopyrimidin-1(2H)-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@@H]1[C@H](O[C@H]([C@@H]1O[Si](C)(C(C)(C)C)C)N2C(N=CC=C2)=O)COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OCCC#N)C(C)C)C

Catalog No. 50-236-4673 Supplier STA PHARMACEUTICAL US LLC Supplier No. STAAM10664KA601GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2’-OTBS Pyrimidinone amidite is a Amidite reagent (Subcategory: Specialty Base Modification) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 155831-90-6
- MDL: MFCD08062082
- InChIKey: UEUDLVPTCOPGMZ-MGVQLQBBSA-N
- Molecular Weight: 845.061761998
- Molecular Formula: C45H61N4O8PSi
- Purity: ≥95%
- Container Type: 60 mL Glass (20-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 63 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3R,4R,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-((tert-butyldimethylsilyl)oxy)-5-(2-oxopyrimidin-1(2H)-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@@H]1[C@H](O[C@H]([C@@H]1O[Si](C)(C(C)(C)C)C)N2C(N=CC=C2)=O)COC(C3=CC=C(C=C3)OC)(C4=CC=C(C=C4)OC)C5=CC=CC=C5)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.