Learn More
TERRISS INDUSTRIES INC Certified Control Drink Flask/Dropper 2 oz One Line
Catalog No. NC0460409
Encompass
These Control Drink Flasks are designed to equate, in terms of capacity, an empty, dry container. The flasks, when properly filled, will hold the exact same content as a bottle and can be used to check proportioning.Calibrated in 1/10 or 1 ml Divisions.
Catalog No. NC0460409 Supplier TERRISS INDUSTRIES INC Supplier No. T09103
These Control Drink Flasks are designed to equate, in terms of capacity, an empty, dry container. The flasks, when properly filled, will hold the exact same content as a bottle and can be used to check proportioning.Calibrated in 1/10 or 1 ml Divisions.