Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Oxa-5-azaspiro[3.5]nonane oxalate, 96%, Thermo Scientific™

Catalog No. AAH57584MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57584MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH57584MD Supplier Thermo Scientific Chemicals Supplier No. H57584MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an pharmaceutical, organic and OLED intermediate.

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1706462-77-2
Molecular Formula C9H15NO5
Quantity 250mg
MDL Number MFCD13180592
Synonym 2-oxa-5-azaspiro 3.5 nonane oxalate
InChI Key OQWJBTNQYYQHRU-UHFFFAOYSA-N
Molecular Weight (g/mol) 217.22
PubChem CID 72207565
Formula Weight 203.19
Percent Purity 96%
Chemical Name or Material 2-Oxa-5-azaspiro[3.5]nonane oxalate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.